COM448635857 Combi-Blocks, Inc.

5,5'-Dicyano-3,3'-bipyridine

  • MDL number MFCD16295131
  • Cas Number : 1226808-65-6
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7346

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7346
Availability details for
Quantity
  • Availability
  • Add to Cart