COM448635865 Combi-Blocks, Inc.

8-Bromo-6-trifluoromethoxyquinoline

  • MDL number MFCD09972204
  • Cas Number : 1020253-25-1
  • Molecular Weight : 292.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7354

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7354
Availability details for
Quantity
  • Availability
  • Add to Cart