COM448635871 Combi-Blocks, Inc.

Dimethyl 3,3'-bipyridine-6,6'-dicarboxylate

  • MDL number MFCD10699711
  • Cas Number : 1033202-47-9
  • Molecular Weight : 272.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7362

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7362
Availability details for
Quantity
  • Availability
  • Add to Cart