COM448635876 Combi-Blocks, Inc.

3-Chloro-2-(4-methoxybenzyloxy)pyridine

  • MDL number MFCD10699712
  • Cas Number : 1033202-56-0
  • Molecular Weight : 249.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7369

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7369
Availability details for
Quantity
  • Availability
  • Add to Cart