COM448635887 Combi-Blocks, Inc.

2-(4-Methoxybenzyloxy)-5-(trifluoromethyl)pyridine

  • MDL number MFCD10699714
  • Cas Number : 1033202-62-8
  • Molecular Weight : 283.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7382

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7382
Availability details for
Quantity
  • Availability
  • Add to Cart