COM448635923 Combi-Blocks, Inc.

3-Amino-2-chloro-6-methoxypyridine, HCl

  • MDL number MFCD11504978
  • Cas Number : 1216499-22-7
  • Molecular Weight : 195.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7726

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7726
Availability details for
Quantity
  • Availability
  • Add to Cart