COM448635925 Combi-Blocks, Inc.

5-Bromo-3-nitropyridine-2-thiol

  • MDL number MFCD07375012
  • Cas Number : 886372-86-7
  • Molecular Weight : 235.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7730

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7730
Availability details for
Quantity
  • Availability
  • Add to Cart