COM448635932 Combi-Blocks, Inc.

N,N-Dimethyl 5-bromopyridine-3-sulfonamide

  • MDL number MFCD11053841
  • Cas Number : 896160-99-9
  • Molecular Weight : 265.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7737

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7737
Availability details for
Quantity
  • Availability
  • Add to Cart