COM448635937 Combi-Blocks, Inc.

2-Acetamido-5-fluoropyridine

  • MDL number MFCD05662406
  • Cas Number : 100304-88-9
  • Molecular Weight : 154.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7743

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7743
Availability details for
Quantity
  • Availability
  • Add to Cart