COM448635942 Combi-Blocks, Inc.

N-Methyl 5-bromopyridine-3-sulfonamide

  • MDL number MFCD11053860
  • Cas Number : 173999-19-4
  • Molecular Weight : 251.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7751

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7751
Availability details for
Quantity
  • Availability
  • Add to Cart