COM448635958 Combi-Blocks, Inc.

5-Methylpyrazine-2-carbohydrazide

  • MDL number MFCD09742108
  • Cas Number : 37545-33-8
  • Molecular Weight : 152.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7775

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7775
Availability details for
Quantity
  • Availability
  • Add to Cart