COM448635959 Combi-Blocks, Inc.

Benzyl 4-chloropyridin-2-ylcarbamate

  • MDL number MFCD11504979
  • Cas Number : 1073372-14-1
  • Molecular Weight : 262.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7778

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7778
Availability details for
Quantity
  • Availability
  • Add to Cart