COM448635962 Combi-Blocks, Inc.

2-Acetamido-5-amino-3-bromopyridine

  • MDL number MFCD11053878
  • Cas Number : 896161-09-4
  • Molecular Weight : 230.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7781

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7781
Availability details for
Quantity
  • Availability
  • Add to Cart