COM448635968 Combi-Blocks, Inc.

Benzyl 5-bromopyridin-3-ylcarbamate

  • MDL number MFCD11504980
  • Cas Number : 762298-10-2
  • Molecular Weight : 307.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7787

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7787
Availability details for
Quantity
  • Availability
  • Add to Cart