COM448635992 Combi-Blocks, Inc.

6-Hydroxy-5-nitronicotinonitrile

  • MDL number MFCD11504985
  • Cas Number : 320405-84-3
  • Molecular Weight : 165.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7821

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7821
Availability details for
Quantity
  • Availability
  • Add to Cart