COM448636020 Combi-Blocks, Inc.

N-(6-Bromopyridin-2-yl)-N-methylacetamide

  • MDL number MFCD11855992
  • Cas Number : 1133116-43-4
  • Molecular Weight : 229.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7862

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7862
Availability details for
Quantity
  • Availability
  • Add to Cart