COM448636039 Combi-Blocks, Inc.

8-Bromo-6-fluoro-4-hydroxyquinoline-3-carboxylic acid

  • MDL number MFCD09861322
  • Cas Number : 1019016-15-9
  • Molecular Weight : 286.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7891

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7891
Availability details for
Quantity
  • Availability
  • Add to Cart