COM448636042 Combi-Blocks, Inc.

N,N-diethylpyridine-4-carboxamide

  • MDL number MFCD00059785
  • Cas Number : 530-40-5
  • Molecular Weight : 178.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7894

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7894
Availability details for
Quantity
  • Availability
  • Add to Cart