COM448636048 Combi-Blocks, Inc.

2-(N'-Acetylhydrazino)-5-chloro-3-fluoropyridine

  • MDL number MFCD12026107
  • Cas Number : 1150561-82-2
  • Molecular Weight : 203.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7903

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7903
Availability details for
Quantity
  • Availability
  • Add to Cart