COM448636071 Combi-Blocks, Inc.

2-(N'-Acetylhydrazino)-6-chloropyridine

  • MDL number MFCD02187335
  • Cas Number : 66999-51-7
  • Molecular Weight : 185.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7932

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7932
Availability details for
Quantity
  • Availability
  • Add to Cart