COM448636074 Combi-Blocks, Inc.

5-Bromo-2-(4-methoxybenzyloxy)pyrimidine

  • MDL number MFCD12026110
  • Cas Number : 1159000-88-0
  • Molecular Weight : 295.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7938

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7938
Availability details for
Quantity
  • Availability
  • Add to Cart