COM448636079 Combi-Blocks, Inc.

3-Bromo-2-pyrrolidinopyridine

  • MDL number MFCD12026111
  • Cas Number : 1150561-84-4
  • Molecular Weight : 227.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7944

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7944
Availability details for
Quantity
  • Availability
  • Add to Cart