COM448636085 Combi-Blocks, Inc.

6-Methoxynicotinamide

  • MDL number MFCD00229166
  • Cas Number : 7150-23-4
  • Molecular Weight : 152.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7951

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7951
Availability details for
Quantity
  • Availability
  • Add to Cart