COM448636100 Combi-Blocks, Inc.

2-Chloro-3-methylpyridine 1-oxide

  • MDL number MFCD14582951
  • Cas Number : 91668-83-6
  • Molecular Weight : 143.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7972

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7972
Availability details for
Quantity
  • Availability
  • Add to Cart