COM448636101 Combi-Blocks, Inc.

2-Chloro-3-methyl-4-nitropyridine 1-oxide

  • MDL number MFCD01194031
  • Cas Number : 60323-95-7
  • Molecular Weight : 188.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7979

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7979
Availability details for
Quantity
  • Availability
  • Add to Cart