COM448636108 Combi-Blocks, Inc.

2-(Cyclohexylamino)nicotinonitrile

  • MDL number MFCD09426708
  • Cas Number : 77276-34-7
  • Molecular Weight : 201.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7986

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7986
Availability details for
Quantity
  • Availability
  • Add to Cart