COM448636114 Combi-Blocks, Inc.

2-Butylamino-5-bromopyridine

  • MDL number MFCD11123176
  • Cas Number : 280116-80-5
  • Molecular Weight : 229.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7993

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7993
Availability details for
Quantity
  • Availability
  • Add to Cart