COM448636115 Combi-Blocks, Inc.

2-Cyclohexylamino-5-bromopyridine

  • MDL number MFCD11124008
  • Cas Number : 942050-72-8
  • Molecular Weight : 255.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7994

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7994
Availability details for
Quantity
  • Availability
  • Add to Cart