COM448636116 Combi-Blocks, Inc.

2-Benzylamino-5-bromopyridine

  • MDL number MFCD04215647
  • Cas Number : 280116-83-8
  • Molecular Weight : 263.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7995

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7995
Availability details for
Quantity
  • Availability
  • Add to Cart