COM448636119 Combi-Blocks, Inc.

5-Bromo-2-ethylaminopyrimidine

  • MDL number MFCD12546487
  • Cas Number : 4214-67-9
  • Molecular Weight : 202.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7998

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-7998
Availability details for
Quantity
  • Availability
  • Add to Cart