COM448636121 Combi-Blocks, Inc.

5-Bromo-2-(2-hydroxyethylamino)pyrimidine

  • MDL number MFCD12546488
  • Cas Number : 1187386-42-0
  • Molecular Weight : 218.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-8000

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-8000
Availability details for
Quantity
  • Availability
  • Add to Cart