COM448636129 Combi-Blocks, Inc.

5-Bromo-2-cyclohexylaminopyrimidine

  • MDL number MFCD03646031
  • Cas Number : 886366-17-2
  • Molecular Weight : 256.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-8008

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-8008
Availability details for
Quantity
  • Availability
  • Add to Cart