COM448636133 Combi-Blocks, Inc.

Ethyl 2-(5-bromopyridin-2-ylidene)-2-cyanoacetate

  • MDL number MFCD00559709
  • Cas Number : 103590-10-9
  • Molecular Weight : 269.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-8014

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-8014
Availability details for
Quantity
  • Availability
  • Add to Cart