COM448636139 Combi-Blocks, Inc.

1-Methyl-4-(3-methylpyridin-2-yl)piperazine

  • MDL number MFCD12546501
  • Cas Number : 1187386-43-1
  • Molecular Weight : 191.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-8023

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-8023
Availability details for
Quantity
  • Availability
  • Add to Cart