COM448636144 Combi-Blocks, Inc.

2-Acetamido-3-picoline

  • MDL number MFCD00495347
  • Cas Number : 7463-30-1
  • Molecular Weight : 150.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-8032

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-8032
Availability details for
Quantity
  • Availability
  • Add to Cart