COM448636147 Combi-Blocks, Inc.

3-Cbz-Amino-2,6-dichloro-5-fluoropyridine

  • MDL number MFCD12913996
  • Cas Number : 1187385-97-2
  • Molecular Weight : 315.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PY-8037

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PY-8037
Availability details for
Quantity
  • Availability
  • Add to Cart