COM448636165 Combi-Blocks, Inc.

2-(2-(4-(Trifluoromethyl)-2-oxopyrimidin-1(2h)-yl)ethyl)isoindoline-1,3-dione

  • MDL number MFCD14585272
  • Cas Number : 1206970-29-7
  • Molecular Weight : 337.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0009

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0009
Availability details for
Quantity
  • Availability
  • Add to Cart