COM448636231 Combi-Blocks, Inc.

tert-Butyl 6-amino-7-chloro-3,4-dihydro-1h-isoquinoline-2-carboxylate

  • MDL number MFCD18157615
  • Cas Number : 912846-75-4
  • Molecular Weight : 282.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0092

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0092
Availability details for
Quantity
  • Availability
  • Add to Cart