COM448636269 Combi-Blocks, Inc.

Ethyl 4-amino-3,5,6-trichloropicolinate

  • MDL number MFCD00183796
  • Cas Number : 91867-42-4
  • Molecular Weight : 269.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0136

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0136
Availability details for
Quantity
  • Availability
  • Add to Cart