COM448636297 Combi-Blocks, Inc.

N-Acetyl-dl-homoserine

  • MDL number MFCD09261290
  • Molecular Weight : 161.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0170
Availability details for
Quantity
  • Availability
  • Add to Cart