COM448636349 Combi-Blocks, Inc.

1-N-(Propan-2-yl)benzene-1,2-diamine

  • MDL number MFCD11186906
  • Cas Number : 70918-95-5
  • Molecular Weight : 150.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0228

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0228
Availability details for
Quantity
  • Availability
  • Add to Cart