COM448636351 Combi-Blocks, Inc.

Methyl 3-amino-3-phenylacrylate

  • MDL number MFCD22381985
  • Cas Number : 70272-01-4
  • Molecular Weight : 177.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0230

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0230
Availability details for
Quantity
  • Availability
  • Add to Cart