COM448636394 Combi-Blocks, Inc.

3-(2-Chlorophenoxy)propan-1-amine HCl

  • MDL number MFCD11036187
  • Cas Number : 50911-59-6
  • Molecular Weight : 222.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0277

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0277
Availability details for
Quantity
  • Availability
  • Add to Cart