COM448636434 Combi-Blocks, Inc.

(S)-4,4-Difluoropyrrolidine-2-carboxylic acid 2,2,2-trifluoroacetic acid

  • MDL number MFCD22194317
  • Cas Number : 1373255-09-4
  • Molecular Weight : 265.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0321

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0321
Availability details for
Quantity
  • Availability
  • Add to Cart