COM448636437 Combi-Blocks, Inc.

4-Anilino-2-(methylthio)pyrimidine-5-carboxylic acid

  • MDL number MFCD11573853
  • Cas Number : 857200-26-1
  • Molecular Weight : 261.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0325

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0325
Availability details for
Quantity
  • Availability
  • Add to Cart