COM448636450 Combi-Blocks, Inc.

tert-Butyl 4-bromophenethyl(methyl)carbamate

  • MDL number MFCD22381983
  • Cas Number : 1191063-30-5
  • Molecular Weight : 314.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0339

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0339
Availability details for
Quantity
  • Availability
  • Add to Cart