COM448636454 Combi-Blocks, Inc.

4-Oxo-piperidine-1,3-dicarboxylic acid 1-benzyl ester 3-ethyl ester

  • MDL number MFCD06738732
  • Cas Number : 154548-45-5
  • Molecular Weight : 305.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0343

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0343
Availability details for
Quantity
  • Availability
  • Add to Cart