COM448636486 Combi-Blocks, Inc.

7-Amino-5-chloro-2(3h)-benzoxazolone

  • MDL number MFCD12024823
  • Cas Number : 889884-60-0
  • Molecular Weight : 184.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0377

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0377
Availability details for
Quantity
  • Availability
  • Add to Cart