COM448636496 Combi-Blocks, Inc.

4-Bromo-5-(3-isocyanatophenyl)-1-methyl-1h-pyrazole

  • MDL number MFCD14585413
  • Cas Number : 1206969-04-1
  • Molecular Weight : 278.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0389

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0389
Availability details for
Quantity
  • Availability
  • Add to Cart