COM448636533 Combi-Blocks, Inc.

N-Boc-beta-alanine-beta-4'-methoxyphenyl-n-carboxyanhydride

  • MDL number MFCD03839896
  • Cas Number : 886362-61-4
  • Molecular Weight : 321.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0436

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QA-0436
Availability details for
Quantity
  • Availability
  • Add to Cart